missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae Chitin deacetylase 2 Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Chitin deacetylase 2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP9473-ye-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16058391

  • 1141.00€ / 500 microgramos


Fecha estimada de envío
23-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_013411.1
S. cerevisiae Chitin deacetylase 2 Protein
500 μg
Not determined.
Full Length
S. cerevisiae
His
Tris-based buffer, 50% glycerol
851017
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CDA2
Recombinant Protein
Yeast
EANREDLKQI DFQFPVLERA ATKTPFPDWL SAFTGLKEWP GLDPPYIPLD FIDFSQIPDY KEYDQNHCDS VPRDSCSFDC HHCTEHDDVY TCSKLSQTFD DGPSASTTKL LDRLKHNSTF FNLGVNIVQH PDIYQRMQKE GHLIGSHTWS HVYLPNVSNE KIIAQIEWSI WAMNATGNHT PKWFRPPYGG IDNRVRAITR QFGLQAVLWD HDTFDWSLLL NDSVITEQEI LQNVINWNKS GTGLILEHDS TEKTVDLAIK INKLIGDDQS TVSHCVGGID YIKEFLS
Videos
Documentos