missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus LDH-A / LDHA Protein
A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) LDH-A / LDHA, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7260-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
WP_001031880.1 | |
S. aureus LDH-A / LDHA Protein | |
Research Use Only | |
LDHA | |
Recombinant Protein | |
E. coli | |
MNKFKGNKVV LIGNGAVGSS YAFSLVNQSI VDELVIIDLD TEKVRGDVMD LKHATPYSPT TVRVKAGEYS DCHDADLVVI CAGAAQKPGE TRLDLVSKNL KIFKSIVGEV MASKFDGIFL VATNPVDILA YATWKFSGLP KERVIGSGTI LDSARFRLLL SEAFDVAPRS VDAQIIGEHG DTELPVWSHA NIAGQPLKTL LEQRPEGKAQ IEQIFVQTRD AAYDIIQAKG ATYYGVAMGL ARITEAIFRN EDAVLTVSAL LEGEYDEEDV YIGVPAVINR NGIRNVVEIP LNDEEQSKFA HSAKTLKDIM AEAEELK | |
Greater than 90% as determined by SDS-PAGE. |
31213039 | |
50 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |