missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Alpha-hemolysin Protein
A DNA sequence encoding the Staphylococcus aureus (strain NCTC 8325) Alpha-hemolysin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7634-ye-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
WP_000857483.1 | |
S. aureus Alpha-hemolysin Protein | |
Research Use Only | |
hly | |
Recombinant Protein | |
Yeast | |
ADSDINIKTG TTDIGSNTTV KTGDLVTYDK ENGMHKKVFY SFIDDKNHNK KLLVIRTKGT IAGQYRVYSE EGANKSGLAW PSAFKVQLQL PDNEVAQISD YYPRNSIDTK EYMSTLTYGF NGNVTGDDTG KIGGLIGANV SIGHTLKYVQ PDFKTILESP TDKKVGWKVI FNNMVNQNWG PYDRDSWNPV YGNQLFMKTR NGSMKAADNF LDPNKASSLL SSGFSPDFAT VITMDRKASK QQTNIDVIYE RVRDDYQLHW TSTNWKGTNT KDKWIDRSSE RYKIDWEKEE MTN | |
Greater than 90% as determined by SDS-PAGE. |
3920722 | |
10 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |