missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rhesus Macaque Procalcitonin Rhesus Protein
A cDNA sequence encoding the Procalcitonin Rhesus was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13156-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Procalcitonin Rhesus Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASAPFRSALESS PDPATLSEEE ARLLLAALVQ DYVQMKASEL EQEQETEGSS LDSPRSKRCG NLSTCMLGTY TQDFNKFHTF PQTAIGVGAP GKKRDMSSDL ERNRRRYVSM PQDAN | |
Greater than 95.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rhesus Macaque | |
His | |
Procalcitonin was filtered (0.4microm) and lyophilized from 0.5 mg/ml solution in 20mM Tris buffer and 50mM NaCl, pH 7.5. |