missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Steroid 17-alpha-hydroxylase / 17,20 lyase Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Steroid 17-alpha-hydroxylase/17,20 lyase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5906-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_036885.1 | |
Steroid 17-alpha-hydroxylase / 17,20 lyase Protein | |
Research Use Only | |
CYP17A1 | |
Recombinant Protein | |
E. coli | |
MWELVGLLLL ILAYFFWVKS KTPGAKLPRS LPSLPLVGSL PFLPRRGHMH VNFFKLQEKY GPIYSLRLGT TTTVIIGHYQ LAREVLIKKG KEFSGRPQMV TQSLLSDQGK GVAFADAGSS WHLHRKLVFS TFSLFKDGQK LEKLICQEAK SLCDMMLAHD KESIDLSTPI FMSVTNIICA ICFNISYEKN DPKLTAIKTF TEGIVDATGD RNLVDIFPWL TIFPNKGLEV IKGYAKVRNE VLTGIFEKCR EKFDSQSISS LTDILIQAKM NSDNNNSCEG RDPDVFSDRH ILATVGDIFG AGIETTTTVL KWILAFLVHN PEVKKKIQKE IDQYVGFSRT PTFNDRSHLL MLEATIREVL RIRPVAPMLI PHKANVDSSI GEFTVPKDTH VVVNLWALHH DENEWDQPDQ FMPERFLDPT GSHLITPTQS YLPFGAGPRS CIGEALARQE LFVFTALLLQ RFDLDVSDDK QLPRLEGDPK VVFLIDPFKV KITVRQAWMD AQAEVST | |
Greater than 90% as determined by SDS-PAGE. |
25146 | |
50 μg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |