missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Sortilin Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Sortilin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6716-ye-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_113955.1 | |
Sortilin Protein | |
Research Use Only | |
Sort1 | |
Recombinant Protein | |
Yeast | |
CEENDYTTWL AHSTDPGDYK DGCILGYKEQ FLRLRKSSVC QNGRDYVVAK QPSICPCSLE DFLCDFGYFR PENASECVEQ PELKGHELEF CLYGKEEHLT TNGYRKIPGD RCQGGMNPAR EVKDLKKKCT SNFLNPKKQN SKSSS | |
Greater than 90% as determined by SDS-PAGE. |
83576 | |
10 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |