missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat MIF Protein
A cDNA sequence encoding the MIF was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12696-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
MIF Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MPMFIVNTNV PRASVPEGFL SELTQQLAQA TGKPAQYIAV HVVPDQLMTF SGTSDPCALC SLHSIGKIGG AQNRNYSKLL CGLLSDRLHI SPDRVYINYY DMNAANVGWN GSTFA | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4 and 5% trehalose. |