missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IL 17A/F Protein

A cDNA sequence encoding the IL 17A/F was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10696-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15994678

Cantidad Precio
1 129.74€ / 2 microgramos
10 121.33€ / 2 microgramos
Envío estimado: 01-03-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.



Descripción

Descripción

Especificaciones

Especificaciones

IL 17A/F Protein
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
IL-17A: MAVLIPQSSV CPNAEANNFL QNVKVNLKVL NSLSSKASSR RPSDYLNRST SPWTLSRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPEKCPF TFRVEKMLVG VGCTCVSSIV RHAS IL-17F: MARRNPKVGL SALQKAGNCP PLEDNSVRVD IRIFNQNQGI SVPRDFQNRS SSPWDYNITR DPDRFPSEIA EAQCRHSGCI NAQGQEDGSM NSVPIQQEIL VLRREPQGCS NSFRLEKMLI KVGCTCVTPI VHHAA
Greater than 98.0% as determined by HPLC and SDS-PAGE.
Research Use Only
The ED50 as determined by its ability to induce of IL-6 production from NIH-3T3 cells, is 15.3-23 ng/ml.
Rat
Untagged
Lyophilized from a concentrated (1 mg/ml) solution containing no additives.
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales