missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IL 12 Protein

A cDNA sequence encoding the IL 12 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10690-0.1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15994658

Cantidad Precio
1 1427.18€ / 100 microgramos
3 1340.41€ / 100 microgramos
Envío estimado: 04-03-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.



Descripción

Descripción

Especificaciones

Especificaciones

IL 12 Protein
100 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Baculovirus
The protein (1 mg/ml) was lyophilized with no additives.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
ED50 range = 0.01-0.05 ng/ml corresponding to a specific activity of 20,000,000-100,000,000IU/mg as determined by the dose dependent secretion of IFN-gamma from PHA and IL-2 activated mouse splenocytes that have been costimulated with PMA. Optimal concentration for individual application should be determined by a dose response assay.
Rat
Untagged
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales