missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat GHBP Protein
A cDNA sequence encoding the GHBP was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11967-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
GHBP Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
FPGSGATPAT LGKASPVLQR INPSLRESSS GKPRFTKCRS PELETFSCYW TEGDDHNLKV PGSIQLYYAR RIAHEWTPEW KECPDYVSAG ANSCYFNSSY TSIWIPYCIK LTTNGDLLKE KCFTVDEIVQ PDPPIGLNWT LLNISLPGIR GDIQVSWQPP PSADVLKGWI ILEYEIQYKE VNETKWKTMS PIWSTSVPLY SLRLDKEHEV RVRSRQRSFE KYSEFSEVLR VTFPQMDTLA ACEEDFRLEH HHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
His | |
GHBP protein solution (0.5 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |