missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Fibroblast growth factor 23 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Fibroblast growth factor 23, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6045-ye-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_570110.1 | |
Fibroblast growth factor 23 Protein | |
Research Use Only | |
Fgf23 | |
Recombinant Protein | |
Yeast | |
YSDTSPLLGS NWGSLTHLYT ATARNSYHLQ IHRDGHVDGT PHQTIYSALM ITSEDAGSVV IIGAMTRRFL CMDLRGNIFG SYHFSPENCR FRQWTLENGY DVYLSPKHHY LVSLGRSKRI FQPGTNPPPF SQFLARRNEV PLLHFYTARP RRHTRSAEDP PERDPLNVLK PRPRATPIPV SCSRELPSAE EGGPAASDPL GVLRRGRGDA RRGAGGTDRC RPFPRFV | |
Greater than 80% as determined by SDS-PAGE. |
170583 | |
200 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |