missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CXCL10 / Crg-2 Protein
A DNA sequence encoding the Rat CXCL10 / Crg-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10286-ec-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_620789.1 | |
CXCL10 / Crg-2 Protein | |
Research Use Only | |
CXCL10 | |
The biological activity determined by a chemotaxis bioassay using human CXCR3 transfected HEK293 cells is in a concentration range of 10-50 ng/ml. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
245920 | |
25 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
IPLARTVRCT CIDFHEQPLR PRAIGKLEII PASLSCPHVE IIATMKKNNE KRCLNPESEA IKSLLKAVSQ RRSKRAP | |
Greater than 95% as determined by SDS-PAGE and HPLC. |