missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CST3 Protein
A cDNA sequence encoding the CST3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11540-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CST3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASGTSRPP PRLLGAPQEA DASEEGVQRA LDFAVSEYNK GSNDAYHSRA IQVVRARKQLVAGINYYLDV EMGRTTCTKS QTNLTNCPFH DQPHLMRKAL CSFQIYSVPW KGTHTLTKSS | |
Greater than 96% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
His | |
Filtered (0.4 micron) and lyophilized from 0.5 mg/ml in 0.03M Acetate buffer, pH 4. |