missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Apolipoprotein C-III Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Apolipoprotein C-III, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8991-ye-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Apolipoprotein C-III Protein | |
Research Use Only | |
APOC3 | |
Recombinant Protein | |
Yeast | |
DEGEGSLLLG SMQGYMEQAS KTVQDALSSM QESDIAVVAS RGWMDNRFKS LKGYWSKFTD KFTGLWESGP EDQLTTPTLE P | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |