missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant RABV NucleoProtein

A DNA sequence encoding the Rabies virus (strain PM) (RABV) Nucleoprotein, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7060-ye-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16057280

  • 2060.00€ / 500 microgramos


Fecha estimada de envío
21-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

RABV NucleoProtein
500 μg
Not determined.
Full Length
RABV
His
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
N
Recombinant Protein
Yeast
MDADKIVFKV NNQVVSLKPE IIVDQYEYKY PAIKDLKKPC ITLGKAPDLN KAYKSVLSGM NAAKLDPDDV CSYLAAAMQF FEGTCPEDWT SYGILIARKG DRITPNSLVE IKRTDVEGNW ALTGGMELTR DPTVSEHASL VGLLLSLYRL SKISGQNTGN YKTNIADRIE QIFETAPFVK IVEHHTLMTT HKMCANWSTI PNFRFLAGTY DMFFSRIEHL YSAIRVGTVV TAYEDCSGLV SFTGFIKQIN LTAREAILYF FHKNFEEEIR RMFEPGQETA VPHSYFIHFR SLGLSGKSPY SSNAVGHVFN LIHFVGCYMG QVRSLNATVI AACAPHEMSV LGGYLGEEFF GKGTFERRFF RDEKELQEYE AAELTKTDVA LADDGTVNSD DEDYFSGETR SPEAVYTRIM MNGGRLKRSH IRRYVSVSSN HQARPNSFAE FLNKTYSNDS
Videos
Documentos