missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit HINT1 Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) HINT1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10110-ma-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001076092.1 | |
HINT1 Protein | |
Research Use Only | |
HINT1 | |
Recombinant Protein | |
Mammal | |
ADEIAKAQVA RPGGDTIFGK IIRKEIPAKI IFEDDQCLAF HDISPQAPTH FLVIPKKHIS QISAAEDADE SLLGHLMIVG KKCAADLGLK KGYRMVVNEG SDGGQSVYHV HLHVLGGRQM NWPPG | |
Greater than 80% as determined by SDS-PAGE. |
100009302 | |
20 μg | |
Not determined. | |
Full Length | |
Rabbit | |
His | |
Tris-based buffer, 50% glycerol |