missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit ALDOB / Aldolase B Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) ALDOB / Aldolase B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5646-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001164585.1 | |
ALDOB / Aldolase B Protein | |
Research Use Only | |
ALDOB | |
Recombinant Protein | |
E. coli | |
AHRFPALTPE QKKELSDIAQ RIVANGKGIL AADESVGTMG NRLQRIKVEN TEENRRQFRE ILFTVDNSIN QSIGGVILFH ETLYQKDSQG KLFRNILKEK GIVVGIKLDQ GGAPLAGTNK ETTIQGLDGL SERCAQYKKD GVDFGKWRAV LRIADQCPSS LAIQENANTL ARYASICQQN GLVPIVEPEV IPDGDHDLEH CQYVTEKVLA AVYKALNDHH VYLEGTLLKP NMVTAGHACT KKYTPEQVAM ATVTALHRTV PAAVPGICFL SGGMSEEDAT LNLNAINLCP LPKPWKLSFS YGRALQASAL AAWGGKAENK KATQEAFMKR AVVNCQAAKG QYVHTGSSGA ASTQSLFTAS YTY | |
Greater than 90% as determined by SDS-PAGE. |
100328925 | |
10 μg | |
Not determined. | |
Full Length | |
Rabbit | |
His-SUMO | |
Tris-based buffer, 50% glycerol |