missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pufferfish Leptin Protein
A cDNA sequence encoding the Leptin was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10749-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Leptin Protein | |
Research Use Only | |
Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. The affinity of human leptin receptors is considerably lower campared to mammalian leptins. | |
PufferFish | |
Untagged | |
The Pufferfish Leptin was lyophilized from a concentrated (0.85 mg/ml) solution with 0.003mM NaHCO3. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MDHILALVLA LLPLSLCVAL PGALDAMDVE KMKSKVTWKA QGLVARIDKH FPDRGLRFDT DKVEGSTSVV ASLESYNNLI SDRFGGVSQI KTEISSLAGY LNHWREGNCQ EQQPKVWPRR NIFNHTVSLE ALMRVREFLK LLQKNVDLLE RC | |
Greater than 99.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |