Learn More
enQuireBio™ Recombinant Pig Parathyroid Hormone Protein
A DNA sequence encoding the Sus scrofa (Pig) Parathyroid Hormone, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10097-ma-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_999547.1 | |
Parathyroid Hormone Protein | |
Research Use Only | |
PTH | |
Recombinant Protein | |
Mammal | |
MTKEEQIFLL HRAQAQCEKR LKEVLQRPAD IMESDKGWAS APTSGKPRKE KASGKLYPES GEDTGSRHQG RPCLPEWDHI LCWP | |
Greater than 90% as determined by SDS-PAGE. |
397675 | |
50 μg | |
Not determined. | |
Full Length | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |