Learn More
enQuireBio™ Recombinant Pig Parathyroid Hormone Protein
A DNA sequence encoding the Sus scrofa (Pig) Parathyroid Hormone, was expressed in the hosts and tags indicated.
Marca:  enQuireBio™ QP10097-ma-100ug
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15962848
  • 1690.00€ / 100 microgramos
Fecha estimada de envío: 11-07-2024
para ver el stock
Especificaciones
NP_999547.1
Parathyroid Hormone Protein
Research Use Only
PTH
Recombinant Protein
Mammal
MTKEEQIFLL HRAQAQCEKR LKEVLQRPAD IMESDKGWAS APTSGKPRKE KASGKLYPES GEDTGSRHQG RPCLPEWDHI LCWP
Greater than 90% as determined by SDS-PAGE.
397675
100 μg
Not determined.
Full Length
Pig
His
Tris-based buffer, 50% glycerol