missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig IL-8 / CXCL8 Protein
Recombinant Protein
Marca: enQuireBio™ QP5522-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_999032.1 | |
IL-8 / CXCL8 Protein | |
Research Use Only | |
CXCL8 | |
Pig | |
Untagged | |
5 mM sodium phosphate, 20 mM sodium chloride, pH 7.5 |
396880 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
ARVSAELRCQ CINTHSTPFH PKFIKELRVI ESGPHCENSE IIVKLVNGKE VCLDPKEKWV QKVVQIFLKR TEKQQQQQ | |
Greater than 95% as determined by SDS-PAGE. |