missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig beta-NGF / Beta-NGF Protein
A DNA sequence encoding the Sus scrofa (Pig) beta-NGF / Beta-NGF Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7637-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
beta-NGF / Beta-NGF Protein | |
Research Use Only | |
Ngf | |
Recombinant Protein | |
E. coli | |
SSSHPVFHRG EFSVCDSVSV WVGDKTTATD IKGKEVMVLG EVNINNSVFK QYFFETKCRD PNPVDSGCRG IDSKHWNSYC TTTHTFVKAL TMDGKQAAWR FIRIDTACVC VLSRKAGRRA | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
Pig | |
His-SUMO | |
Tris-based buffer, 50% glycerol |