missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ovine GH Antagonist Protein
A cDNA sequence encoding the GH Antagonist was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10640-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
GH Antagonist Protein | |
Research Use Only | |
GH G119R acts as antagonist using an in vitro bioassay in PDF-P1 3B9 cells stably transfected with rabbit GH receptors. It is capable of forming a 1:1 complex with the recombinant ovine growth hormone receptor extracellular domain (ECD) and binds to this ECD with affinity similar the the wild type oGH. | |
Ovine | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |