missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other TBEV gE C-end Protein
A cDNA sequence encoding the TBEV gE C-end was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13699-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TBEV gE C-end Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |
Encephalitis protein is >95% pure as determined by 10% PAGE (coomassie staining). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
20mM MES pH 6.5, 8M urea, 200mM NaCl and 0.05% Tween-20. |