missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other T. pallidum p47 (Partial) Protein
A cDNA sequence encoding the T. pallidum p47 (Partial) was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13676-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
T. pallidum p47 (Partial) Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
70mM Tris-HCl pH 8.0, 50mM NaCl, 50% Glycerol and 1.5M urea. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Treponema Pallidum protein is >95% pure as determined by 10% PAGE (coomassie staining). |