missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Streptavidin-NC Protein
A cDNA sequence encoding the Streptavidin-NC was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13627-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Streptavidin-NC Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The sterile solution contains 10mM K2HPO4-KH2PO4, pH 7.3. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 94.0% as determined by SDS-PAGE. |