missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other ProMatrilysin Protein
A cDNA sequence encoding the ProMatrilysin was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10852-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
ProMatrilysin Protein | |
Research Use Only | |
The specific activity was found to be 1400 IU/mg. | |
Other | |
Untagged | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The protein contains the following additives 25mM Tris-HCl (pH 7.5),150mM NaCl, 5mM CaCl2, 0.01% Brij-35 and 0.02% NaN3. |