missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other MIP2 Viral Protein
A cDNA sequence encoding the MIP2 Viral was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10788-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
MIP2 Viral Protein | |
Research Use Only | |
Determined by the inhibitory effect on monocyte migration response to human MIP1A using a concentration range of 1microg-10microg/ml of viral MIP2 which will inhibit 25ng/ml of human MIP1A. | |
Other | |
Untagged | |
Lyophilized from 1 mg/ml solution containing 20mM Phospahte Buffer, pH 7.4 % 0.15M NaCl. |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
LGASWHRPDK CCLGYQKRPL PQVLLSSWYP TSQLCSKPGV IFLTKRGRQVCADKSKDWVK KLMQQLPVTA | |
Greater than 97.0% as determined by RP-HPLC & SDS-PAGE. |