missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other GLB1 Protein
A cDNA sequence encoding the GLB1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11980-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
945006 | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MTMITDSLAV VLQRRDWENP GVTQLNRLAA HPPFASWRNS EEARTDRPSQ QLRSLNGEWR FAWFPAPEAV PESWLECDLP EADTVVVPSN WQMHGYDAPI YTNVTYPITV NPPFVPTENP TGCYSLTFNV DESWLQEGQT RIIFDGVNSA FHLWCNGRWV GYGQDSRLPS EFDLSAFLRA GENRLAVMVL RWSDGSYLED QDMWRMSGIF RDVSLLHKPT TQISDFHVAT RFNDDFSRAV LEAEVQMCGE LRDYLRVTVS LWQGETQVAS GTAPFGGEII DERGGYADRV TLRLNVENPK LWSAEIPNLY RAVVELHTAD GTLIEAEACD VGFREVRIEN GLLLLNGKPL LIRGVNRHEH HPLHGQVMDE QTMVQDILLM KQNNFNAVRC SHYPNHPLWY TLCDRYGLYV VDEANIETHG MVPMNRLTDD PRWLPAMSER VTRMVQRDRN HPSVIIWSLG NESGHGANHD ALYRWIKSVD PSRPVQYEGG GADTTATDII CPMYARVDED QPFPAVPKWS IKKWLSLPGE TRPLILCEYA HAMGNSLGGF AKYWQAFRQY PRLQGGFVWD WVDQSLIKYD ENGNPWSAYG GDFGDTPNDR QFCMNGLVFA DRTPHPALTE AKHQQQFFQF RLSGQTIEVT SEYLFRHSDN ELLHWMVALD GKPLASGEVP LDVAPQGKQL IELPELPQPE SAGQLWLTVR VVQPNATAWS EAGHISAWQQ WRLAENLSVT LPAASHAIPH LTTSEMDFCI ELGNKRWQFN RQSGFLSQMW IGDKKQLLTP LRDQFTRAPL DNDIGVSEAT RIDPNAWVER WKAAGHYQAE AALLQCTADT LADAVLITTA HAWQHQGKTL FISRKTYRID GSGQMAITVD VEVASDTPHP ARIGLNCQLA QVAERVNWLG LGPQENYPDR | |
Protein is >95% pure as determined by SDS-PAGE, by measuring optical density at 280 nm and by method of Bradford et al. |
GLB1 Protein | |
Research Use Only | |
lacZ | |
Other | |
Untagged | |
Beta-Galactosidase (1 mg/1ml) is formulated in 8M urea, 20mM Tris-HCl pH 8.0, and 10mM beta-mercaptoethanol |