missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other CAS9 S. Pyogenes Protein
A cDNA sequence encoding the CAS9 S. Pyogenes was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11279-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CAS9 S. Pyogenes Protein | |
Research Use Only | |
The incubation of 20pmol of CAS9 protein with 1microg pUC19 supercoiled vector at 37C for 4 hours leads to formation of less than 10% open circular pUC19 form as determined by gel electrophoresis. Also observed 97% of PCR product digestion after 30 minutes at 37C. | |
Other | |
His | |
The CAS9 solution contains 10mM Tris-HCl, 300mM NaCl, 0.1mM EDTA, 50% glycerol and 1mM DTT pH 7.5. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MDKKYSIGLD IGTNSVGWAV ITDEYKVPSK KFKVLGNTDR HSIKKNLIGA LLFDSGETAE ATRLKRTARR RYTRRKNRIC YLQEIFSNEM AKVDDSFFHR LEESFLVEED KKHERHPIFG NIVDEVAYHE KYPTIYHLRK KLVDSTDKAD LRLIYLALAH MIKFRGHFLI EGDLNPDNSD VDKLFIQLVQ TYNQLFEENP INASGVDAKA ILSARLSKSR RLENLIAQLP GEKKNGLFGN LIALSLGLTP NFKSNFDLAE DAKLQLSKDT YDDDLDNLLA QIGDQYADLF LAAKNLSDAI LLSDILRVNT EITKAPLSAS MIKRYDEHHQ DLTLLKALVR QQLPEKYKEI FFDQSKNGYA GYIDGGASQE EFYKFIKPIL EKMDGTEELL VKLNREDLLR KQRTFDNGSI PHQIHLGELH AILRRQEDFY PFLKDNREKI EKILTFRIPY YVGPLARGNS RFAWMTRKSE ETITPWNFEE VVDKGASAQS FIERMTNFDK NLPNEKVLPK HSLLYEYFTV YNELTKVKYV TEGMRKPAFL SGEQKKAIVD LLFKTNRKVT VKQLKEDYFK KIECFDSVEI SGVEDRFNAS LGTYHDLLKI IKDKDFLDNE ENEDILEDIV LTLTLFEDRE MIEERLKTYA HLFDDKVMKQ LKRRRYTGWG RLSRKLINGI RDKQSGKTIL DFLKSDGFAN RNFMQLIHDD SLTFKEDIQK AQVSGQGDSL HEHIANLAGS PAIKKGILQT VKVVDELVKV MGRHKPENIV IEMARENQTT QKGQKNSRER MKRIEEGIKE LGSQILKEHP VENTQLQNEK LYLYYLQNGR DMYVDQELDI NRLSDYDVDH IVPQSFLKDD SIDNKVLTRS DKNRGKSDNV PSEEVVKKMK NYWRQLLNAK LITQRKFDNL TKAERGGLSE | |
Greater than 90% as determined by SDS-PAGE. |