missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other a-Actinin Protein
A cDNA sequence encoding the a-Actinin was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10923-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
a-Actinin Protein | |
Research Use Only | |
Recombinant Protein | |
Non-species Specific | |
MDHHYDPQQT NDYMQPEEDW DRDLLLDPAW EKQQRKTFTA WCNSHLRKAG TQIENIEEDF RDGLKLMLLL EVISGERLAK PERGKMRVHK ISNVNKALDF IASKGVKLVS IGAEEIVDGN VKMTLGMIWT IILRFAIQDI SVEETSAKEG LLLWCQRKTA PYKNVNIQNF HISWKDGLGF CALIHRHRPE LIDYGKLRKD DPLTNLNTAF DVAEKYLDIP KMLDAEDIVG TARPDEKAIM TYVSSFYHAF SGAQKAETAA NRICKVLAVN QENEQLMEDY EKLASDLLEW IRRTIPWLEN RAPENTMQAM QQKLEDFRDY RRLHKPPKVQ EKCQLEINFN TLQTKLRLSN RPAFMPSEGK MVSDINNAWG GLEQAEKGYE EWLLNEIRRL ERLDHLAEKF RQKASIHESW TDGKEAMLQQ KDYETATLSE IKALLKKHEA FESDLAAHQD RVEQIAAIAQ ELNELDYYDS PSVNARCQKI CDQWDNLGAL TQKRREALER TEKLLETIDQ LYLEYAKRAA PFNNWMEGAM EDLQDTFIVH TIEEIQGLTT AHEQFKATLP DADKERQAIL GIHNEVSKIV QTYHVNMAGT NPYTTITPQE INGKWEHVRQ LVPRRDQALM EEHARQQQNE RLRKQFGAQA NVIGPWIQTK MEEIGRISIE MHGTLEDQLN HLRQYEKSIV NYKPKIDQLE GDHQQIQEAL IFDNKHTNYT MEHIRVGWEQ LLTTIARTIN EVENQILTRD AKGISQEQMN EFRASFNHFD RDHSGTLGPE EFKACLISLG YDIGNDAQGE AEFARIMSIV DPNRMGVVTF QAFIDFMSRE TADTDTADQV MASFKILAGD KNYITVDELR RELPPDQAEY CIARMAPYNG RDAVPGALDY MSFSTALYGE SDL | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The protein was lyophilized from a 1 mg/ml solution containing 10mM Tris acetate buffer pH 7.6, 0.1mM EDTA, 2mM DTT, and 20mM NaCl. |