Learn More
enQuireBio™ Recombinant Mycoplasma Probable cytosol aminopeptidase Protein
A DNA sequence encoding the Mycoplasma pneumoniae (strain ATCC 29342 / M129) Probable cytosol aminopeptidase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6948-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_110261.1 | |
Mycoplasma Probable cytosol aminopeptidase Protein | |
Research Use Only | |
pepA | |
Recombinant Protein | |
E. coli | |
MKLNKLFDSK TVVFKKSDKY GKSDCCKTKC VEGQIEFGVK IPTDFPAFNR ALVTFLKTQK NQLNVDLDSF VELYKAENLC YKTALKVVVA SITFCETTPF TMKTEPQKNV EVAVKCDSEH TSLIKEYEVV GNYVNMARQL QDTPSDQLYP EEFVKRFEKA ATGLGVKITV LKQADLIKKK MGLLLGVNKG SEREARLLVI SYNNNKKSSE TLALVGKGIT YDSGGMNIKT GDYMRGMKYD MSGAAIVCST VLALAKNKVK TNVVAVAALT ENLPGPHAQR PDDIQTAYNG KTVEIDNTDA EGRLVLADAI SYAAKDLKAT QIIDVATLTG LMSYILSTTY TGIFSTCDMA WDAFKKAACC AGEPVWRLPM HPDYLKPLES KLADLQNSTS VKGAGSSRAA CFLAEFREGV PLIHCDIAST ASIQDLGQGV LVRTLYERAA QQAKE | |
Greater than 90% as determined by SDS-PAGE. |
876829 | |
500 μg | |
Not determined. | |
Full Length | |
Mycoplasma | |
His-SUMO | |
Tris-based buffer, 50% glycerol |