missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Thrombospondin-3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Thrombospondin-3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6787-ye-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_038719.2 | |
Thrombospondin-3 Protein | |
Research Use Only | |
Thbs3 | |
Recombinant Protein | |
Yeast | |
DLQVIDLLTV GESRQMVAVA EKIRTALLTA GDIYLLSTFR LPPKQGGVLF GLYSRQDNTR WLEASVVGKI NKVLVRYQRE DGKVHAVNLQ QAGLADGRTH TALLRLRGPS RPSPGLQLYV DCKLGDQHAG LPALAPIPPA EVSGLEIRTG QKAYLRMQGF VESMKIILGG SMARVGALSE CPFQGDDSIH NAVTSALQSI LGEQTKALVT QLTLFNQILV ELRDDIRDQV KEMSLIRNTI MECQVCGFHE QRSHCSPSPC FRGVDCMEVY EYPGYRCGPC PPGLQGNGTH CDDINECAHA DPCFPGSSCI NTMPGFHCEA CPPGYKG | |
Greater than 90% as determined by SDS-PAGE. |
21827 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |