Learn More
enQuireBio™ Recombinant Mouse SHH / Sonic hedgehog Protein
Recombinant Protein
Marca: enQuireBio™ QP5463-500ug
Detalles adicionales : Peso : 0.01000kg
Specifications
NP_033196.1 | |
SHH / Sonic hedgehog Protein | |
Research Use Only | |
SHH | |
Recombinant Protein | |
E. coli | |
MIIGPGRGFG KRRHPKKLTP LAYKQFIPNV AEKTLGASGR YEGKITRNSE RFKELTPNYN PDIIFKDEEN TGADRLMTQR CKDKLNALAI SVMNQWPGVK LRVTEGWDED GHHSEESLHY EGRAVDITTS DRDRSKYGML ARLAVEAGFD WVYYESKAHI HCSVKAENSV AAKSGG | |
Greater than 95% as determined by SDS-PAGE. |
20423 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Sonic Hedgehog ( SHH ) activity was confirmed in a CCL-226 cell proliferatin assay. See figure for results. | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |