missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Serum amyloid A-2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Serum amyloid A-2 protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6649-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_035444.1 | |
Serum amyloid A-2 Protein | |
Research Use Only | |
Saa2 | |
Recombinant Protein | |
E. coli | |
GFFSFIGEAF QGAGDMWRAY TDMKEAGWKD GDKYFHARGN YDAAQRGPGG VWAAEKISDA RESFQEFFGR GHEDTMADQE ANRHGRSGKD PNYYRPPGLP AKY | |
Greater than 90% as determined by SDS-PAGE. |
20209 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |