missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Serine protease inhibitor Kazal-type 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Serine protease inhibitor Kazal-type 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7376-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_033284.1 | |
Serine protease inhibitor Kazal-type 1 Protein | |
Research Use Only | |
Spink3 | |
Recombinant Protein | |
E. coli | |
AKVTGKEASC HDAVAGCPRI YDPVCGTDGI TYANECVLCF ENRKRIEPVL IRKGGPC | |
Greater than 80% as determined by SDS-PAGE. |
20730 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |