missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Placenta-expressed transcript 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Placenta-expressed transcript 1 protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7974-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_083915.2 | |
Placenta-expressed transcript 1 Protein | |
Research Use Only | |
Plet1 | |
Recombinant Protein | |
E. coli | |
SDNGSCVVLD NIYTSDILEI STMANVSGGD VTYTVTVPVN DSVSAVILKA VKEDDSPVGT WSGTYEKCND SSVYYNLTSQ SQSVFQTNWT VPTSEDVTKV NLQVLIVVNR TASKSSVKME QVQPSASTPI PESSETSQTI NTTPTVNTAK TTAKDTANTT AVTTANTTAN TTAVTTAKTT AKSLAIRTLG S | |
Greater than 86.6% as determined by SDS-PAGE. |
76509 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |