missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Nuclear respiratory factor 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Nuclear respiratory factor 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6435-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001157698.1 | |
Nuclear respiratory factor 1 Protein | |
Research Use Only | |
Nrf1 | |
Recombinant Protein | |
E. coli | |
MEEHGVTQTE HMATIEAHAV AQQVQQVHVA TYTEHSMLSA DEDSPSSPED TSYDDSDILN STAADEVTAH LAAAGPVGMA AAAAVATGKK RKRPHVFESN PSIRKRQQTR LLRKLRATLD EYTTRVGQQA IVLCISPSKP NPVFKVFGAA PLENVVRKYK SMILEDLESA LAEHAPAPQE VNSELPPLTI DGIPVSVDKM TQAQLRAFIP EMLKYSTGRG KPGWGKESCK PIWWPEDIPW ANVRSDVRTE EQKQRVSWTQ ALRTIVKNCY KQHGREDLLY AFEDQQTQTQ ATTTHSIAHL VPSQTVVQTF SNPDGTVSLI QVGTGATVAT LADASELPTT VTVAQVNYSA VADGEVEQNW ATLQGGEMTI QTTQASEATM S-QAVASLAE AAVAASQEMQ QGATVTMALN SEAAAHAVAT LAEATLQGGG QIVLSGETAA AVGALTGVQD ANGLVQIPVS MYQTVVTSLA QGNGPVQVAM APVTTRISDS AVTMDGQAVE VVTLEQ | |
Greater than 90% as determined by SDS-PAGE. |
18181 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |