missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Nqo2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Nqo2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8176-ye-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001156713.1 | |
Nqo2 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
18105 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Nqo2 | |
Recombinant Protein | |
Yeast | |
MAGKKVLIVY AHQEPKSFNG SLKKVAVEEL SKQGCTVTVS DLYSMNFEPR ATRNDITGAP SNPDVFSYGI ETHEAYKKKA LTSDIFEEQR KVQEADLVIF QFPLYWFSVP AILKGWMDRV LCRGFAFDIP GFYDSGFLKG KLALLSLTTG GTAEMYTKDG VSGDFRYFLW PLQHGTLHFC GFKVLAPQIS FGLDVSSEEE RKVMLASWAQ RLKSIWKEEP IHCTPPWYFQ E |