missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Nidogen-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Nidogen-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6420-ye-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_035047.2 | |
Nidogen-1 Protein | |
Research Use Only | |
Nid1 | |
Recombinant Protein | |
Yeast | |
SPQRVNGKVK GRIFVGSSQV PVVFENTDLH SYVVMNHGRS YTAISTIPET VGYSLLPLAP IGGIIGWMFA VEQDGFKNGF SITGGEFTRQ AEVTFLGHPG KLVLKQQFSG IDEHGHLTIS TELEGRVPQI PYGASVHIEP YTELYHYSSS VITSSSTREY TVMEPDQDGA APSHTHIYQW RQTITFQECA HDDARPALPS TQQLSVDSVF VLYNKEERIL RYALSNSIGP VRDGSPDA | |
Greater than 90% as determined by SDS-PAGE. |
18073 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |