missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Neurofilament Protein
A DNA sequence encoding the Mus musculus (Mouse) Neurofilament, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9303-ye-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_035040.1 | |
Neurofilament Protein | |
Research Use Only | |
NEFL | |
Recombinant Protein | |
Yeast | |
SSFGYDPYFS TSYKRRYVET PRVHISSVRS GYSTARSAYS SYSAPVSSSL SVRRSYSSSS GSLMPSLENL DLSQVAAISN DLKSIRTQEK AQLQDLNDRF ASFIERVHEL EQQNKVLEAE LLVLRQKHSE PSRFRALYEQ EIRDLRLAAE DATNEKQALQ GEREGLEETL RNLQARYEEE VLSREDAEGR LMEARKGADE AALARAELEK RIDSLMDEIA FLKKVHEEEI AELQAQIQYA QISVEMDVSS KPDLSAALKD IRAQYEKLAA KNMQNAEEWF KSRFTVLTES AAKNTDAVRA AKDEVSESRR LLKAKTLEIE ACRGMNEALE KQLQELEDKQ NADISAMQDT INKLENELRS TKSEMARYLK EYQDLLNVKM ALDIEIAAYR KLLEGEETRL SFTSVGSITS GYSQSSQVFG RSAYSGLQSS SYLMSARSFP AYYTSHVQEE QTEVEETIEA TKAEEAKDEP PSEGEAEEEE KEKEEGEEEE GAEEEEAAKD ESEDTKEEEE GGEGEEEDTK ESEEEEKKEE SAGEEQVAKK KD | |
Greater than 90% as determined by SDS-PAGE. |
18039 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |