missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MCP-3 / CCL7 Protein
A DNA sequence encoding the Mouse MCP-3 / CCL7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10267-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_038682.1 | |
MCP-3 / CCL7 Protein | |
Research Use Only | |
CCL7 | |
The biologically active determined by a chemotaxis bioassay using human monocytes is in a concentration range of 100-300 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
20306 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
QPDGPNASTC CYVKKQKIPK RNLKSYRRIT SSRCPWEAVI FKTKKGMEVC AEAHQKWVEE AIAYLDMKTP TPKP | |
Greater than 95% as determined by SDS-PAGE and HPLC. |