missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Inactive tyrosine-protein kinase 7 Protein
A DNA sequence encoding the Mus musculus (Mouse) Inactive tyrosine-protein kinase 7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9829-ye-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_780377.1 | |
Inactive tyrosine-protein kinase 7 Protein | |
Research Use Only | |
Ptk7 | |
Recombinant Protein | |
Yeast | |
AIVFIKEPSS QDALQGRRAL LRCEVEAPDP VHVYWLLNGV PVQDTERRFA QGSSLSFAAV DRLQDSGAFQ CVARDNVTGE EVRSTNASFN IKWIEAGPVV LKHPASEAEI QPQTQVTLRC HIDGHPRPTY QWFRDGTPLS DDQSTHTVSS RERNLTLRPA SPEHSGLYSC CAHNAFGQAC SSQNFTLSVA DESFARVVLA PQDVVVARNE EAMFHCQFSA QPPPSLQWVF EDETPITNRS RPPHLRRAVV FANGSLLLTQ VRPRNAGVYR CIGQGQRGPP IVLEATLHLA EIEDMLPFEP RVFIAGDEER VTCPAPQGLP TPSVWWEHAG VPLPAHGRVH QKGLELVFVT IAESDTGVYT CHASNLAGQR RQDVNITVAT VPTWLRKPQD SQLEEGKPGY LHCLTQATPK PTVIWYRNQM LISEDSRFEV SKNGTLRINS VEVYDGTLYR CVSSTPAGSI EAQARVQVLE KLKFTPPPQP QQCMEFDKEA TVPCSATGRE KPTVKWVRAD GSSLPEWVTD NAGTLHFARV TRDDAGNYTC IASNEPQGQI RAHVQLTVAV FITFKVEPER TTVYQGHTAL LRCEAQGDPK PLIQWKGKDR ILDPTKLGPR MHIFQNGSLV IHDVAPEDSG SYTCIAGNSC NIRHTEAPLL VVDKPVMEDS EGPGSPPPYK MIQT | |
Greater than 80% as determined by SDS-PAGE. |
71461 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |