missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL1F6 / IL36 Protein
A DNA sequence encoding the Mouse IL1F6 / IL36, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10465-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_062323.1 | |
IL1F6 / IL36 Protein | |
Research Use Only | |
Il36a | |
The specific activity determined by its ability in a functional ELISA. Immobilized rMuIL-36alpha at 1 ug/mL can bind recombinant murine IL-1 Rrp2 with a range of 0.15-5 ug/mL. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, 5 % trehalose, lyophilized |
54448 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MNKEKELRAA SPSLRHVQDL SSRVWILQNN ILTAVPRKEQ TVPVTITLLP CQYLDTLETN RGDPTYMGVQ RPMSCLFCTK DGEQPVLQLG EGNIMEMYNK KEPVKASLFY HKKSGTTSTF ESAAFPGWFI AVCSKGSCPL ILTQELGEIF ITDFEMIVVH | |
Greater than 95% as determined by SDS-PAGE and HPLC. |