missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL17F / IL-17F Protein
Recombinant Protein
Marca: enQuireBio™ QP1479-100 ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_665855.2 | |
IL17F / IL-17F Protein | |
Research Use Only | |
IL17F | |
Recombinant Protein | |
E. coli | |
MRKNPKAGVP ALQKAGNCPP LEDNTVRVDI RIFNQNQGIS VPREFQNRSS SPWDYNITRD PHRFPSEIAE AQCRHSGCIN AQGQEDSTMN SVAIQQEILV LRREPQGCSN SFRLEKMLLK VGCTCVKPIV HQAA | |
Greater than 95% as determined by SDS-PAGE. |
257630 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse IL17F / IL-17F induces IL-6 production in 3T3 cells (typical ED50 is ? 1 ug/ml). | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |