missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Hadh, mitochondrial Protein
A DNA sequence encoding the Mus musculus (Mouse) Hadh, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7685-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_032238.2 | |
Hadh, mitochondrial Protein | |
Research Use Only | |
Hadh | |
Recombinant Protein | |
E. coli | |
SSSSSASAAA KKILIKHVTV IGGGLMGAGI AQVAAATGHT VVLVDQTEDI LAKSKKGIEE SLKRMAKKKF TENPKAGDEF VEKTLSCLST STDAASVVHS TDLVVEAIVE NLKLKNELFQ RLDKFAAEHT IFASNTSSLQ ITNIANATTR QDRFAGLHFF NPVPMMKLVE VIKTPMTSQK TFESLVDFCK TLGKHPVSCK DTPGFIVNRL LVPYLIEAVR LHERGDASKE DIDTAMKLGA GYPMGPFELL DYVGLDTTKF ILDGWHEMEP ENPLFQPSPS MNNLVAQKKL GKKTGEGFYK YK | |
Greater than 90% as determined by SDS-PAGE. |
15107 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |