missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Gp2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Gp2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8169-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_080265.2 | |
Gp2 Protein | |
Research Use Only | |
Gp2 | |
Recombinant Protein | |
E. coli | |
STIHQGYGRP RNSSNLDLDC GSPDSPSSGI CFDPCQNHTV LNDPTRSTEN NDSSVAWCDD NLHGWYRFVG DGGVKMPETC VSVFRCHTSA PMWLSGSHPI LGDGIVSHTA CANWNENCCF WRSEVQVKAC SEELGEYHVY KLQGTPECSL RYCTDPSTAP KNCEITCRPE EECVFQNNNW SCVCRQDLHV SDSQSLQPLL DCGDNEIKVK LDKCLLGGMG FKEEIIAYLN DRNCNGTMQD EPNNWVSMTS PVVANYCGNI LEKNGTHAIY RNTLSLATDF IIRDFRVNVN FQCAYPLDMS VSLETALQPI VSSLTVDVDG AGEFNVKMAL FQDQSYTNPY EGAEVLLPVE SILYVGVLLN RGDTSRFKLL LTNCYATPSE DRHDPVKYFI IKNRCPNQRD STINVRENGV SSESRFSVQM FMFAGNYDLV FLHCEVYLCD STTEQCQPSC STNRLRSSRP AIDYNRVLDL GPITKRSAQS SATSKGTPHT TGFLLAWPMF FLPVFLALLF | |
Greater than 90% as determined by SDS-PAGE. |
67133 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |