missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Galectin-7 Protein
A DNA sequence encoding the Mus musculus (Mouse) Galectin-7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6299-ye-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Galectin-7 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
LGALS7 | |
Recombinant Protein | |
Yeast | |
SATHHKTSLP QGVRVGTVMR IRGLVPDQAG RFHVNLLCGE EQGADAALHF NPRLDTSEVV FNTKQQGKWG REERGTGIPF QRGQPFEVLL IATEEGFKAV VGDDEYLHFH HRLPPARVRL VEVGGDVQLH SLNIF |