missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Galectin-4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Galectin-4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9839-ye-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_034836.1 | |
Galectin-4 Protein | |
Research Use Only | |
Lgals4 | |
Recombinant Protein | |
Yeast | |
MAYVPAPGYQ PTYNPTLPYK RPIPGGLSVG MSVYIQGMAK ENMRRFHVNF AVGQDDGADV AFHFNPRFDG WDKVVFNTMQ SGQWGKEEKK KSMPFQKGKH FELVFMVMPE HYKVVVNGNS FYEYGHRLPV QMVTHLQVDG DLELQSINFL GGQPAAAPYP GAMTIPAYPA GSPGYNPPQM NTLPVMTGPP VFNPRVPYVG ALQGGLTVRR TIIIKGYVLP TARNFVINFK VGSSGDIALH LNPRIGDSVV RNSFMNGSWG AEERKVAYNP FGPGQFFDLS IRCGMDRFKV FANGQHLFDF SHRFQAFQMV DTLEINGDIT LSYVQI | |
Greater than 90% as determined by SDS-PAGE. |
16855 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |