missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Frataxin, mitochondrial Protein
A DNA sequence encoding the Mus musculus (Mouse) Frataxin, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6059-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_032070.1 | |
Frataxin, mitochondrial Protein | |
Research Use Only | |
Fxn | |
Recombinant Protein | |
E. coli | |
LGTLDNPSSL DETAYERLAE ETLDSLAEFF EDLADKPYTL EDYDVSFGDG VLTIKLGGDL GTYVINKQTP NKQIWLSSPS SGPKRYDWTG KNWVYSHDGV SLHELLAREL TKALNTKLDL SSLAYSGKGT | |
Greater than 90% as determined by SDS-PAGE. |
14297 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |